2-Bromophenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromophenylhydrazine hydrochloride, 1-(2-Bromophenyl)hydrazine hydrochloride

A683687-25G A683687-1000G A683687-100G A683687-10G A683687-1G A683687-500G A683687-5G
76844-752EA 20 USD
76844-752 77117-844 77117-846 77117-848 77117-850 77117-852 77117-854
2-Bromophenylhydrazinium chloride 98%
2-Bromophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₈BrClN₂
MDL Number: MFCD00012926
CAS Number: 50709-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR