(R)-N-(3-Aminopropyl)-N-(1-(3-benzyl-7-chloro-4-oxo-4H-chromen-2-yl)-2-methylpropyl)-4-methylbenzamide hydrochloride ≥98%

Supplier: Ambeed
Warning

A654863-1MG A654863-50MG A654863-5MG
76844-590EA 50.93 USD
76844-590 76844-592 76844-594 76844-596
(R)-N-(3-Aminopropyl)-N-(1-(3-benzyl-7-chloro-4-oxo-4H-chromen-2-yl)-2-methylpropyl)-4-methylbenzamide hydrochloride ≥98%
(R)-N-(3-Aminopropyl)-N-(1-(3-benzyl-7-chloro-4-oxo-4H-chromen-2-yl)-2-methylpropyl)-4-methylbenzamide hydrochloride

Precaución: for Research Use Only

Formula: C₃₁H₃₄Cl₂N₂O₃
MDL Number: MFCD18385010
CAS Number: 940929-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR