Triphenylphosphonium bromide 97%

Supplier: Ambeed
Danger

Synonyms: Triphenylphosphine hydrobromide

A644739-25G A644739-10G A644739-100G
76844-538EA 7 USD
76844-538 77117-618 77686-598
Triphenylphosphonium bromide 97%
Triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₁₈H₁₆BrP
MDL Number: MFCD00035107
CAS Number: 6399-81-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR