N-Phenylmaleimide 98%

Supplier: Ambeed
Danger

A633542-25G A633542-100G
76844-472EA 6.37 USD
76844-472 77673-722
N-Phenylmaleimide 98%
N-Phenylmaleimide

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD00005502
CAS Number: 941-69-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR