2-Mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A631903-1G A631903-100MG A631903-250MG A631903-5G
76844-444EA 303.42 USD
76844-444 77145-162 77145-164 77145-166
2-Mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate 97%
2-Mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₄H₁₈BF₄N₃
MW: 315.11 g/mol
MDL Number: MFCD28133590
CAS Number: 862893-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR