2',4'-Dihydroxyacetophenone 98%

Supplier: Ambeed
Warning

A552806-25G A552806-1000G A552806-100G A552806-100MG A552806-10G A552806-500G
76843-912EA 11.67 USD
76843-912 77116-916 77116-918 77116-920 77116-922 77116-924
2',4'-Dihydroxyacetophenone 98%
2',4'-Dihydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD00002279
CAS Number: 89-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR