Ethyl bromopyruvate 80%

Supplier: Ambeed
Danger

Synonyms: Bromopyruvic acid ethyl ester, Ethyl 3-bromo-2-oxopropanoate, 3-Bromo-2-oxopropionic acid ethyl ester

A533127-25G A533127-100G A533127-10G A533127-5G
76843-712EA 11.67 USD
76843-712 77169-610 77169-612 77169-614
Ethyl bromopyruvate 80%
Ethyl bromopyruvate

Precaución: for Research Use Only

Formula: BrH₂CC(O)COOCH₂CH₃
MW: 195.01 g/mol
MDL Number: MFCD00000204
CAS Number: 70-23-5
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR