2-Mercaptobenzimidazole 97%

Supplier: Ambeed
Danger

Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

A523734-25G A523734-100G A523734-500G
76843-646EA 5.31 USD
76843-646 77265-936 77265-938
2-Mercaptobenzimidazole 97%
2-Mercaptobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD00466107
CAS Number: 583-39-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR