N-(1',2-Dihydroxy-[1,2'-binaphthalen]-4'-yl)-4-methoxybenzenesulfonamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
F-1113789, F1113-0789, C188-9, SKPin C1
A445130-10MG
A445130-25MG
A445130-50MG
A445130-1MG
A445130-5MG
76842-572EA
82.76
USD
76842-572
76842-574
76842-576
77265-030
77307-340
N-(1',2-Dihydroxy-[1,2'-binaphthalen]-4'-yl)-4-methoxybenzenesulfonamide 98%
N-(1',2-Dihydroxy-[1,2'-binaphthalen]-4'-yl)-4-methoxybenzenesulfonamide
Precaución: for Research Use Only
Formula:
C₂₇H₂₁NO₅S MW: 471.52 g/mol |
MDL Number:
MFCD03141581 CAS Number: 432001-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...