1-Fluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: o-Fluoronitrobenzene, 2-Fluoronitrobenzene

A439205-25G A439205-1000G A439205-100G A439205-10G A439205-500G
76842-462EA 6.37 USD
76842-462 77156-024 77156-026 77156-028 77156-030
1-Fluoro-2-nitrobenzene 98%
1-Fluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
MDL Number: MFCD00007048
CAS Number: 1493-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR