4-(Dimethylamino)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Dimethylaminobenzoic acid

A404890-25G A404890-1000G A404890-100G A404890-10G A404890-500G
76841-638EA 20 USD
76841-638 77033-690 77033-692 77033-694 77033-696
4-(Dimethylamino)benzoic acid 97%
4-(Dimethylamino)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD00002537
CAS Number: 619-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR