Methyl-4-(chloroformyl)benzoate 98%

Supplier: Ambeed
Danger

Synonyms: Terephthalic Acid Monomethyl Ester Chloride, 4-(Chloroformyl)benzoic Acid Methyl Ester, Methyl 4-(chlorocarbonyl)benzoate, Methyl 4-(Chlorocarbonyl)benzoate

A400960-25G A400960-100G A400960-10G A400960-5G
76841-540EA 18.04 USD
76841-540 77264-516 77264-518 77264-520
Methyl-4-(chloroformyl)benzoate 98%
Methyl-4-(chloroformyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MW: 198.60 g/mol
MDL Number: MFCD00059486
CAS Number: 7377-26-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR