1,1-Bis(hydroxymethyl)cyclopropane 98%

Supplier: Ambeed
Warning

A399781-25G A399781-1000G A399781-100G A399781-10G A399781-500G
76841-514EA 8 USD
76841-514 76990-214 76990-216 76990-218 76990-220 76990-222
1,1-Bis(hydroxymethyl)cyclopropane 98%
1,1-Bis(hydroxymethyl)cyclopropane

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD00464207
CAS Number: 39590-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR