Thymidine (2'-deoxythymidine) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
A392461-25G
A392461-1000G
A392461-100G
A392461-10G
A392461-1G
A392461-500G
A392461-5G
76841-340EA
19
USD
76841-340
77278-634
77278-636
77278-638
77278-640
77278-642
77278-644
Thymidine (2'-deoxythymidine) 98%
Thymidine (2'-deoxythymidine)
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₂O₅ |
MDL Number:
MFCD00006537 CAS Number: 50-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...