2,5-Dimethyl-2,5-hexanediol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,1,4,4-Tetramethyl-1,4-butanediol
A381673-25G
A381673-100G
76841-054EA
5.31
USD
76841-054
77003-648
2,5-Dimethyl-2,5-hexanediol 98%
2,5-Dimethyl-2,5-hexanediol
Precaución: for Research Use Only
Formula:
C₈H₁₈O₂ MW: 146.23 g/mol |
MDL Number:
MFCD00004473 CAS Number: 110-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...