1-Bromo-3,5-difluorobenzene 97%

Supplier: Ambeed
Danger

Synonyms: 3,5-Difluorobromobenzene

A349801-25G A349801-100G A349801-10G
76840-120EA 6.37 USD
76840-120 77106-784 77106-786
1-Bromo-3,5-difluorobenzene 97%
1-Bromo-3,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrF₂
MW: 192.99 g/mol
MDL Number: MFCD00010305
CAS Number: 461-96-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR