3-Bromobenzaldehyde 97%

Supplier: Ambeed
Warning

A348729-25G A348729-1000G A348729-100G A348729-10G A348729-500G
76840-062EA 10 USD
76840-062 77106-746 77106-748 77106-750 77106-752
3-Bromobenzaldehyde 97%
3-Bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO
MDL Number: MFCD00003345
CAS Number: 3132-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR