Cytosine 98%

Supplier: Ambeed
Warning

A347875-25G A347875-100G A347875-10G A347875-500G
76840-040EA 6 USD
76840-040 77278-098 77278-100 77278-102
Cytosine 98%
Cytosine

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MDL Number: MFCD00006034
CAS Number: 71-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR