α,α,α-Trifluoro-o-cresol 98%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxybenzotrifluoride, 2-(Trifluoromethyl)phenol

A341531-25G A341531-10G
76839-840EA 12.74 USD
76839-840 76978-412
α,α,α-Trifluoro-o-cresol 98%
α,α,α-Trifluoro-o-cresol

Precaución: For research use only

Formula: C₇H₅F₃O
MW: 162.11 g/mol
MDL Number: MFCD00002222
CAS Number: 444-30-4
UN: 2921
ADR: 8, 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR