2-(2-Hydroxy-5-methylphenyl)benzotriazole ≥99%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Benzotriazolyl)-p-cresol, Drometrizole

A335992-25G A335992-100G A335992-500G A335992-1000G
76839-686EA 6.37 USD
76839-686 77032-988 77032-990 77601-512
2-(2-Hydroxy-5-methylphenyl)benzotriazole ≥99%
2-(2-Hydroxy-5-methylphenyl)benzotriazole

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O
MW: 225.25 g/mol
MDL Number: MFCD00022903
CAS Number: 2440-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR