4-Ethylphenol 98%

Supplier: Ambeed
Danger

A333517-25G A333517-100G A333517-10G A333517-1G A333517-500G A333517-5G A333517-1MG
76839-622EA 10.61 USD
76839-622 77032-946 77032-948 77032-950 77032-952 77032-954 77601-502
4-Ethylphenol 98%
4-Ethylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O
MW: 122.17 g/mol
MDL Number: MFCD00002393
CAS Number: 123-07-9
UN: 2430
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR