Ethyl-6-chloronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-chloronicotinate

A328887-25G A328887-10G A328887-1G A328887-500G A328887-5G
76839-424EA 21.22 USD
76839-424 77239-712 77239-714 77239-716 77239-718
Ethyl-6-chloronicotinate 97%
Ethyl-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD00082739
CAS Number: 49608-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR