6-Bromo-m-toluidine 98%

Supplier: Ambeed
Warning

A311831-25G A311831-100G A311831-10G A311831-1G A311831-5G
76838-680EA 17 USD
76838-680 77167-130 77167-132 77167-134 77167-136
6-Bromo-m-toluidine 98%
6-Bromo-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MDL Number: MFCD01806278
CAS Number: 53078-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR