1,6-Hexanediol 95%

Supplier: Ambeed
Warning

A310995-25G A310995-1000G A310995-100G A310995-500G
76838-616EA 9 USD
76838-616 77194-638 77194-640 77194-642
1,6-Hexanediol 95%
1,6-Hexanediol

Precaución: for Research Use Only

Formula: C₆H₁₄O₂
MDL Number: MFCD00002985
CAS Number: 629-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR