1H,1H,2H,2H-Tridecafluoro-1-octanol 98%

Supplier: Ambeed
Warning

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol

A310731-25G A310731-100G A310731-10G A310731-500G A310731-5G
76838-588EA 13.8 USD
76838-588 77239-426 77239-428 77239-430 77239-432
1H,1H,2H,2H-Tridecafluoro-1-octanol 98%
1H,1H,2H,2H-Tridecafluoro-1-octanol

Precaución: for Research Use Only

Formula: C₈H₅F₁₃O
MW: 364.11 g/mol
MDL Number: MFCD00042143
CAS Number: 647-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR