3-Chloropropyl acetate 98%

Supplier: Ambeed
Warning

A310577-25G A310577-1000G A310577-100G A310577-10G A310577-500G A310577-5G
76838-584EA 8.49 USD
76838-584 77277-512 77277-514 77277-516 77277-518 77277-520
3-Chloropropyl acetate 98%
3-Chloropropyl acetate

Precaución: for Research Use Only

Formula: C₅H₉ClO₂
MW: 136.58 g/mol
MDL Number: MFCD00001000
CAS Number: 628-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR