2-Bromo-m-xylene 98%

Supplier: Ambeed
Warning

A301545-25G A301545-100G A301545-10G A301545-500G
76838-156EA 7 USD
76838-156 77114-482 77114-484 77114-486
2-Bromo-m-xylene 98%
2-Bromo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₉Br
MDL Number: MFCD00000075
CAS Number: 576-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR