N-Methylmorpholine-N-oxide 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide

A299370-25G A299370-1000G A299370-100G A299370-10G A299370-1G A299370-500G
76838-064EA 8.49 USD
76838-064 77289-934 77289-936 77289-938 77289-940 77289-942
N-Methylmorpholine-N-oxide 97%
N-Methylmorpholine-N-oxide

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00005947
CAS Number: 7529-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR