1,4-Diiodobenzene 98%

Supplier: Ambeed
Warning

A296319-25G A296319-100G A296319-10G A296319-500G A296319-5G
76837-896EA 30 USD
76837-896 77114-386 77114-388 77114-390 77114-392
1,4-Diiodobenzene 98%
1,4-Diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285
MDL Number: MFCD00001054
CAS Number: 624-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR