7-Chloroquinolin-4-ol 95%

Supplier: Ambeed
Warning

A293380-25G A293380-10G A293380-500G A293380-5G
76837-734EA 39 USD
76837-734 76875-516 76875-518 77593-794
7-Chloroquinolin-4-ol 95%
7-Chloroquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD00006778
CAS Number: 86-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR