2-Methoxy-p-cresol 98%

Supplier: Ambeed
Warning

A291186-25G A291186-1000G A291186-100G A291186-10G A291186-500G
76837-620EA 17 USD
76837-620 76875-460 76875-462 76875-464 76875-466
2-Methoxy-p-cresol 98%
2-Methoxy-p-cresol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MDL Number: MFCD00002378
CAS Number: 93-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR