5,6,7-Trifluoroindoline-2,3-dione 95%

Supplier: Ambeed
Warning

A999437-100MG A999437-1G A999437-250MG
76833-176EA 35 USD
76833-176 76833-178 76833-180
5,6,7-Trifluoroindoline-2,3-dione 95%
5,6,7-Trifluoroindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₂F₃NO₂
MDL Number: MFCD09739194
CAS Number: 107583-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR