2-Methylimidazo[1,2-a]pyrimidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A999103-100MG A999103-1G A999103-250MG
76833-170EA 63.66 USD
76833-170 76833-172 76833-174
2-Methylimidazo[1,2-a]pyrimidine-3-carboxylic acid 95%
2-Methylimidazo[1,2-a]pyrimidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MDL Number: MFCD07368659
CAS Number: 90830-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR