2-(6-Bromopyridin-2-yl)propan-2-ol 97%

Supplier: Ambeed
Warning

A995634-100MG A995634-1G A995634-250MG A995634-5G
76833-152EA 18 USD
76833-152 76833-154 76833-156 76833-158
2-(6-Bromopyridin-2-yl)propan-2-ol 97%
2-(6-Bromopyridin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD16657880
CAS Number: 638218-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR