2-(4-Bromo-2,6-difluorophenyl)acetonitrile 95%

Supplier: Ambeed
Warning

A995210-250MG A995210-5G A995210-1G
76833-148EA 44 USD
76833-148 76833-150 77886-960
2-(4-Bromo-2,6-difluorophenyl)acetonitrile 95%
2-(4-Bromo-2,6-difluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrF₂N
MDL Number: MFCD03094464
CAS Number: 537033-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR