2-(Propylamino)-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Propyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A991280-1G A991280-250MG A991280-5G
76833-142EA 150.65 USD
76833-142 76833-144 76833-146
2-(Propylamino)-5-pyrimidineboronic acid pinacol ester 98%
2-(Propylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₂BN₃O₂
MW: 263.14 g/mol
MDL Number: MFCD12546557
CAS Number: 1218791-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR