Chlorosulfonyl acetic acid ethyl ester 95%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-(chlorosulphonyl)acetate

A988376-1G A988376-250MG A988376-25G A988376-5G
76833-134EA 24.41 USD
76833-134 76833-136 76833-138 76833-140
Chlorosulfonyl acetic acid ethyl ester 95%
Chlorosulfonyl acetic acid ethyl ester

Precaución: for Research Use Only

Formula: C₄H₇ClO₄S
MW: 186.614 g/mol
MDL Number: MFCD08458866
CAS Number: 55896-93-0
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR