(3-Butoxy-5-methoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A986138-100MG A986138-1G A986138-250MG
76833-126EA 36.08 USD
76833-126 76833-128 76833-130
(3-Butoxy-5-methoxyphenyl)boronic acid 98%
(3-Butoxy-5-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₄
MW: 224.06 g/mol
MDL Number: MFCD17926490
CAS Number: 1256355-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR