1-(4-Bromo-2-methoxyphenyl)ethanone 98%

Supplier: Ambeed
Warning

A984387-100MG A984387-1G A984387-250MG A984387-5G A984387-25G
76833-120EA 14.86 USD
76833-120 76833-122 76833-124 77569-708 77684-915
1-(4-Bromo-2-methoxyphenyl)ethanone 98%
1-(4-Bromo-2-methoxyphenyl)ethanone

Precaución: For research use only.

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD11848490
CAS Number: 89368-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR