(S)-2,4-Diaminobutanoic acid 98%

Supplier: Ambeed
Warning

A983607-1G A983607-25G A983607-5G
76833-114EA 14.86 USD
76833-114 76833-116 76833-118
(S)-2,4-Diaminobutanoic acid 98%
(S)-2,4-Diaminobutanoic acid

Precaución: for Research Use Only

Formula: C₄H₁₀N₂O₂
MW: 118.13 g/mol
MDL Number: MFCD00082632
CAS Number: 1758-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR