2-Chlor-6-methyl-4-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A980214-100MG A980214-250MG A980214-5G
76833-102EA 24 USD
76833-102 76833-104 76833-106
2-Chlor-6-methyl-4-pyridineboronic acid pinacol ester 98%
2-Chlor-6-methyl-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
MDL Number: MFCD08063106
CAS Number: 697739-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR