6-(Methoxycarbonyl)-2-naphthoic acid 97%

Supplier: Ambeed
Warning

A970007-100MG A970007-1G A970007-250MG
76833-094EA 10 USD
76833-094 76833-096 76833-098
6-(Methoxycarbonyl)-2-naphthoic acid 97%
6-(Methoxycarbonyl)-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₄
MW: 230.21 g/mol
MDL Number: MFCD00805122
CAS Number: 7568-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR