5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 5-(Bromomethyl)-2-fluorophenylboronic acid, pinacol ester, 2-(5-(Bromomethyl)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A967877-100MG A967877-1G A967877-250MG A967877-5G
76833-080EA 37 USD
76833-080 76833-082 76833-084 76833-086
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 98%
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MDL Number: MFCD17926519
CAS Number: 1256360-47-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR