(6-Methylquinolin-8-yl)boronic acid 98%

Supplier: Ambeed
Warning

A964523-1G A964523-250MG A964523-5G
76833-070EA 210.06 USD
76833-070 76833-072 76833-074
(6-Methylquinolin-8-yl)boronic acid 98%
(6-Methylquinolin-8-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₂
MW: 187.00 g/mol
MDL Number: MFCD17214249
CAS Number: 1256355-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR