4'-(4-Methylphenyl)-2,2':6',2''-terpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(p-Tolyl)-2,2:6,2-terpyridine

A956950-100MG A956950-1G A956950-250MG A956950-5G
76833-058EA 10 USD
76833-058 76833-060 76833-062 76833-064
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine 95%
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine

Precaución: for Research Use Only

Formula: C₂₂H₁₇N₃
MW: 323.40 g/mol
MDL Number: MFCD06796977
CAS Number: 89972-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR