(3-Methoxy-5-propoxyphenyl)boronic acid 96%

Supplier: Ambeed
Warning

A953265-100MG A953265-1G A953265-250MG
76833-046EA 36.08 USD
76833-046 76833-048 76833-050
(3-Methoxy-5-propoxyphenyl)boronic acid 96%
(3-Methoxy-5-propoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₄
MW: 210.04 g/mol
MDL Number: MFCD17926489
CAS Number: 1256355-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR