2-Chloro-3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 6-Chloro-5-methoxypyridine-3-boronic acid pinacol ester

A951263-100MG A951263-1G A951263-250MG
76833-034EA 35.01 USD
76833-034 76833-036 76833-038
2-Chloro-3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
2-Chloro-3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₃
MW: 269.53 g/mol
MDL Number: MFCD16660309
CAS Number: 1256360-28-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR