4-Amino-3-methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A947476-100MG A947476-1G A947476-250MG
76833-020EA 153.84 USD
76833-020 76833-022 76833-024
4-Amino-3-methyl-1H-pyrazole-5-carboxylic acid 95%
4-Amino-3-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.12 g/mol
MDL Number: MFCD17276602
CAS Number: 94993-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR