5-Nitro-o-anisidine 98%

Supplier: Ambeed
Warning

A944741-100G A944741-5G A944741-25G
76833-016EA 105 USD
76833-016 76833-018 77886-925
5-Nitro-o-anisidine 98%
5-Nitro-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD00007261
CAS Number: 99-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR