1H-Pyrrolo[3,2-c]pyridine-6-carbonitrile 95%

Supplier: Ambeed
Warning

A943258-250MG A943258-1G
76833-014EA 60.48 USD
76833-014 77684-889
1H-Pyrrolo[3,2-c]pyridine-6-carbonitrile 95%
1H-Pyrrolo[3,2-c]pyridine-6-carbonitrile

Precaución: For research use only.

Formula: C₈H₅N₃
MW: 143.15 g/mol
MDL Number: MFCD11845528
CAS Number: 1082040-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR